Tisk…

Dichloro(p-cymene)ruthenium(II) dimer 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

363290010 363290050
ACRO363290010EA 2400 CZK
ACRO363290010 ACRO363290050
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer

CAS No.: 52462-29-0

Vzorec: C₂₀H₂₈Cl₄Ru₂
Storage Temperature: Ambient
MDL Number: MFCD00064793
CAS číslo: 52462-29-0

Objednat

Výsledky specifikačního testu

Appearance Orange-red to red-brown Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR