(R)-2-Isopropylsuccinic acid-1-methyl ester ≥95%, ee 98%
Dodavatel: Thermo Fisher Scientific
363065000
363060050
ACRO363065000EA
6620
CZK
ACRO363065000
ACRO363060050
(R)-2-Isopropylsuccinic acid-1-methyl ester ≥95%, ee 98%
(R)-2-Isopropylsuccinic acid-1-methyl ester
CAS No.: 220498-08-8
Vzorec:
C₈H₁₄O₄ MW: 174.196 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01091030 CAS číslo: 220498-08-8 |
Výsledky specifikačního testu
Appearance | Clear yellow Viscous liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Enantiomeric excess | ≥97.5 % (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...