Tisk…

(Acetylacetonato)(norbornadiene)rhodium(I)

Dodavatel: Thermo Fisher Scientific
Varování

382561000 382565000
ACRO382561000EA 3310 CZK
ACRO382561000 ACRO382565000
(Acetylacetonato)(norbornadiene)rhodium(I)
(Acetylacetonato)(norbornadiene)rhodium(I)

Appearance: Yellow Powder

Vzorec: C₁₂H₁₅O₂Rh
Bod tání: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075051
CAS číslo: 32354-50-0
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Yellow Powder
Infrared spectrum Conforms
Rhodium (Rh) ≥34.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR