Tisk…
Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: Trityltetra(pentafluorophenyl)borate, Triphenylmethylium tetrakis(perfluorophenyl)borate, Trityl tetrakis(pentafluorophenyl)borate, Triphenylcarbenium Tetrakis(pentafluorophenyl)borate, Tritylium Tetrakis(pentafluorophenyl)borate
382390010 382390050
ACRO382390010EA 14120 CZK
ACRO382390010 ACRO382390050
Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Triphenylmethylium tetrakis(pentafluorophenyl)borate
Appearance: Yellow to dark yellow Powder
Vzorec: C₄₃H₁₅BF₂₀
MW: 922.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426981
CAS číslo: 136040-19-2
UN: 2811
ADR: 6.1 III
Objednat
Výsledky specifikačního testu
Appearance Yellow to dark yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %