Tisk…

Benzyl propargyl ether 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: ((Prop-2-yn-1-yloxy)methyl)benzene

380870010 380870050
ACRO380870010EA 938 CZK
ACRO380870010 ACRO380870050
Benzyl propargyl ether 97%
Benzyl propargyl ether

Appearance: Clear colorless to light yellow Liquid

Vzorec: C₁₀H₁₀O
MW: 146,189 g/mol
Bod varu: 100…101 °C (18 mmHg)
Hustota: 1,011 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00483987
CAS číslo: 4039-82-1

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5220 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR