Tisk…

Tris(trimethylsilyl)amine 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Nonamethyltrisilazane, Silanamine,1,1,1-trimethyl-N,N-bis(trimethylsilyl)-, Amine, tris(trimethylsilyl)-, Nitrilotris(trimethylsilane)

378770050 378770250
ACRO378770050EA 1640 CZK
ACRO378770050 ACRO378770250
Tris(trimethylsilyl)amine 99%
Tris(trimethylsilyl)amine

Appearance: Colorless Crystals

Vzorec: C₉H₂₇NSi₃
MW: 233,577 g/mol
Bod varu: 72 °C (10 mmHg)
Bod tání: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00047990
CAS číslo: 1586-73-8
UN: 3259
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance Colorless Crystals
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR