Tisk…

Hydridotetrakis(triphenylphosphine)rhodium(I)

Dodavatel: Thermo Scientific

Synonyms: Tetrakis(triphenylphosphine)rhodium(I) Hydride

377930010 377932500
ACRO377930010EA 7800 CZK
ACRO377930010 ACRO377932500
Hydridotetrakis(triphenylphosphine)rhodium(I)
Hydridotetrakis(triphenylphosphine)rhodium(I)

Appearance: Ochre Crystalline powder

Vzorec: C₇₂H₆₁P₄Rh
MW: 1153,08 g/mol
Bod tání: 140…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00015867
CAS číslo: 18284-36-1

Objednat

Výsledky specifikačního testu

Appearance Ochre Crystalline powder
Infrared spectrum Conforms
Rhodium (Rh) ≥8.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR