Methyl-L-threoninate hydrochloride 98+%
Dodavatel: Thermo Scientific
Synonyms:
(2S,3R)-Methyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OMe·HCl, L-Threonine methyl ester hydrochloride
376690010
376690050
376690250
ACRO376690010EA
1440
CZK
ACRO376690010
ACRO376690050
ACRO376690250
Methyl-L-threoninate hydrochloride 98+%
Methyl-L-threoninate hydrochloride
CAS No.: 39994-75-7
Vzorec:
C₅H₁₂ClNO₃ MW: 169,605 g/mol Bod tání: 64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037677 CAS číslo: 39994-75-7 |
Výsledky specifikačního testu
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.0 % (On dry substance) |
Water | ≤1.0 % |
Specific optical rotation | -9.5° to -10.5° (20°C, 589 nm) (c=1, Methanol, 20°C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...