Tisk…

Methyl-2-(3-fluorophenyl)acetate 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Methyl 2-(3-fluorophenyl)acetate, Methyl 3-Fluorophenylacetate, 3-Fluorophenylacetic acid methyll ester, Methyl 3-fluorophenylacetate

351090050 351090250
ACRO351090050EA 842 CZK
ACRO351090050 ACRO351090250
Methyl-2-(3-fluorophenyl)acetate 98%
Methyl-2-(3-fluorophenyl)acetate

Appearance: Clear colorless to light yellow Liquid

Vzorec: C₉H₉FO₂
MW: 168.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093288
CAS číslo: 64123-77-9

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4890 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR