2,2-Di(tert-butylperoxy)butane 50% in aromatic free mineral spirit
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
349830100
349835000
ACRO349830100EA
611
CZK
ACRO349830100
ACRO349835000
2,2-Di(tert-butylperoxy)butane 50% in aromatic free mineral spirit
2,2-Di(tert-butylperoxy)butane
Appearance: Clear solution
Vzorec:
C₁₂H₂₆O₄ Hustota: 0,805 g/cm³ Bod vzplanutí: 43 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00048243 CAS číslo: 2167-23-9 UN: 3103 ADR: 5.2, |
Výsledky specifikačního testu
Appearance | Clear solution |
Titration Iodometric | 49 to 51 % |
Color scale | ≤10 APHA |
Impurity | ≤0.3 % Hydroperoxides (as TBHP) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...