Tisk…
Ethynylmagnesium chloride solution 0.6 M in THF/toluene, AcroSeal®
Dodavatel: Thermo Scientific
Nebezpečí
388951000 388958000
388951000.EA 2330 CZK
388951000. 388958000.
Ethynylmagnesium chloride solution 0.6 M in THF/toluene, AcroSeal®
Ethynylmagnesium chloride solution
Appearance: Yellow to dark brown Solution
Vzorec: C₂HClMg
MW: 84,785 g/mol
Hustota: 0,96 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00075343
CAS číslo: 65032-27-1
UN: 3399
ADR: 4.3,I
Objednat
Výsledky specifikačního testu
Appearance Yellow to dark brown Solution
Titration Grignard 4 to 6 % (As ethynyl-MgCl)