Tisk…

1-(2-Chloroethyl)pyrrolidine hydrochloride 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: N-(2-Chloroethyl)pyrrolidine hydrochloride

109220250 109221000
ACRO109220250EA 1580 CZK
ACRO109220250 ACRO109221000
1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
1-(2-Chloroethyl)pyrrolidine hydrochloride

Appearance: White to off-white Crystalline powder or crystals

Vzorec: C₆H₁₂ClN
MW: 170,08 g/mol
Bod tání: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012718
CAS číslo: 7250-67-1
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 165 to 175 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR