Tisk…

Methyl-2-(m-tolyl)acetate 98%

Dodavatel: Thermo Scientific

Synonyms: Methyl 3-methylphenylacetate

374080050 374080250
ACRO374080050EA 1300 CZK
ACRO374080050 ACRO374080250
Methyl-2-(m-tolyl)acetate 98%
Methyl-2-(m-tolyl)acetate

Appearance: Clear colorless to light yellow Liquid

Vzorec: C₁₀H₁₂O₂
MW: 164,204 g/mol
Bod varu: 117 °C (13 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00968468
CAS číslo: 53088-69-0

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5045 to 1.5065 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR