Amberlite® IRA-402 (Cl) ion-exchange resin
Dodavatel: Thermo Fisher Scientific
369942500
369940010
369940050
ACRO369942500EA
1250
CZK
ACRO369942500
ACRO369940010
ACRO369940050
Amberlite® IRA-402 (Cl) ion-exchange resin
Amberlite® IRA-402 (Cl)
CAS No.: 52439-77-7
Storage Temperature:
Ambient |
MDL Number:
MFCD00145564 CAS číslo: 52439-77-7 |
Výsledky specifikačního testu
Appearance | Light yellow Semitransparent beads |
Particle size | ≤5.0 % (> 1180 micron) |
Harmonic mean size | 0.600 to 0.750 mm |
Uniformity coefficient | ≤1.60 |
Moisture holding capacity | 49.0 to 60.0 % (Cl) |
Total volume capacity | ≥1.20 eq/l (Cl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...