Tisk…

(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

367750010 367750050
ACRO367750010EA 1760 CZK
ACRO367750010 ACRO367750050
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 99%
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine

Appearance: Clear colorless to light yellow Liquid

Vzorec: C₇H₁₆N₂
Hustota: 0,919 g/cm³
Bod vzplanutí: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00191371
CAS číslo: 22795-99-9
UN: 2734
ADR: 8,I

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4660 to 1.4680 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -105° to -109° (20°C, 589 nm) (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR