1-[2-(Morpholin-4-yl)ethyl]piperazine 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-[2-(Piperazin-1-yl)ethyl]morpholine, 1-[2-(Morpholin-4-yl)-ethyl]-piperazine
367140050
367140010
ACRO367140050EA
10590
CZK
ACRO367140050
ACRO367140010
1-[2-(Morpholin-4-yl)ethyl]piperazine 99%
1-[2-(Morpholin-4-yl)ethyl]piperazine
Appearance: White to light yellow Low melting crystals or viscous liquid
Vzorec:
C₁₀H₂₁N₃O MW: 199,298 g/mol Bod varu: 82…84 °C (,5mbar) Storage Temperature: Ambient |
MDL Number:
MFCD00191217 CAS číslo: 4892-89-1 UN: 2735 ADR: 8,III |
Výsledky specifikačního testu
Appearance | White to light yellow Low melting crystals or viscous liquid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...