Tisk…

Rhodamine B base 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Xanthylium, 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-, inner salt, 9-(2-Carboxylatophenyl)-3,6-bis(diethylamino)xanthylium, Ethaminium, N-(9-(2-carboxyphenyl)-6-(diethylamino)-3H-xanthen-3-ylidene)-N-ethyl)-, hydroxide, inner salt

215230250 215231000
ACRO215230250EA 2940 CZK
ACRO215230250 ACRO215231000
Rhodamine B base 97%
Rhodamine B base

CAS No.: 509-34-2

Vzorec: C₂₈H₃₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00066967
CAS číslo: 509-34-2
UN: 3077

Objednat

Výsledky specifikačního testu

Appearance Pink to red to violet Powder and/or chunks
Infrared spectrum Conforms
Dye content ≥96.0 %
UV Lambda max 542 to 546 nm (in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR