Tisk…

Iron(III) hexacyanoferrate(II), pure

Dodavatel: Thermo Fisher Scientific

Synonyms: Iron(III) Ferrocyanide, Prussian Blue

215210250 215211000
ACRO215210250EA 1980 CZK
ACRO215210250 ACRO215211000
Iron(III) hexacyanoferrate(II), pure
Iron(III) hexacyanoferrate(II)

CAS No.: 14038-43-8

Vzorec: C₁₈Fe₇N₁₈
MW: 859.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135663
CAS číslo: 14038-43-8

Objednat

Výsledky specifikačního testu

Appearance Dark blue Powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR