Indigotrisulphonic acid potassium salt
Dodavatel: Thermo Scientific
|
Varování
|
215170010
215170050
215170250
ACRO215170010EA
1620
CZK
ACRO215170010
ACRO215170050
ACRO215170250
Indigotrisulphonic acid potassium salt
Indigotrisulphonic acid potassium salt
CAS No.: 67627-18-3
Vzorec:
C₁₆H₇K₃N₂O₁₁S₃ MW: 616,71 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00013160 CAS číslo: 67627-18-3 |
Výsledky specifikačního testu
Appearance | Dark blue Crystalline powder |
Infrared spectrum | Conforms |
Molar extinction coefficient | ≥ 17000 (250 - 256 nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...