Methyl-2-amino-3-hydroxypropanoate hydrochloride 99%
Dodavatel: Thermo Fisher Scientific
207450100
207451000
ACRO207450100EA
1340
CZK
ACRO207450100
ACRO207451000
Methyl-2-amino-3-hydroxypropanoate hydrochloride 99%
Methyl-2-amino-3-hydroxypropanoate hydrochloride
CAS No.: 5619-04-5
Vzorec:
C₄H₁₀ClNO₃ Bod tání: 130…135 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012593 CAS číslo: 5619-04-5 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...