Tisk…

Allylpalladium(II) chloride dimer 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer

206835000 206830050
ACRO206835000EA 3410 CZK
ACRO206835000 ACRO206830050
Allylpalladium(II) chloride dimer 98%
Allylpalladium(II) chloride dimer

CAS No.: 12012-95-2

Vzorec: C₆H₁₀Cl₂Pd₂
MW: 365,89 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00044874
CAS číslo: 12012-95-2

Objednat

Výsledky specifikačního testu

Appearance Yellow to yellow-green Crystalline powder or crystals
Assay ≥56.7 % (Pd)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR