Tisk…

Potassium tri-sec-butylborohydride 1 M in THF, AcroSeal®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: K-Selectride

205471000 205478000
205471000.EA 6260 CZK
205471000. 205478000.
Potassium tri-sec-butylborohydride 1 M in THF, AcroSeal®
Potassium tri-sec-butylborohydride

Appearance: Clear light yellow Solution

Vzorec: C₁₂H₂₇BK
MW: 221.256 g/mol
Hustota: 0.91 g/cm³
Bod vzplanutí: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00009324
CAS číslo: 54575-49-4
UN: 3399
ADR: 4.3 I

Objednat

Výsledky specifikačního testu

Appearance Clear light yellow Solution (May darken on storage)
Strength ≥1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR