Tisk…
DL-Isocitric acid, trisodium salt hydrate 95%
Dodavatel: Thermo Fisher Scientific
205010010 205010050 205010250
ACRO205010010EA 2410 CZK
ACRO205010010 ACRO205010050 ACRO205010250
DL-Isocitric acid, trisodium salt hydrate 95%
DL-Isocitric acid, trisodium salt hydrate
Appearance: White to off-white Crystalline powder
Vzorec: C₆H₅Na₃O₇·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149284
CAS číslo: 1637-73-6
Objednat
Výsledky specifikačního testu
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤20 % (K.F.)
Titration with HClO4 ≥94 % (On dry substance)