Tisk…

Sodium glycinate 99%

Dodavatel: Thermo Fisher Scientific

204591000 204595000
ACRO204591000EA 2580 CZK
ACRO204591000 ACRO204595000
Sodium glycinate 99%
Sodium glycinate

CAS No.: 6000-44-8

Vzorec: C₂H₄NNaO₂
Bod tání: 197…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00062543
CAS číslo: 6000-44-8
EINECS: 227-842-3

Objednat

Výsledky specifikačního testu

Appearance Colorless to white to off-white Powder or crystals
Infrared spectrum Conforms
Water ≤1.5 % (K.F.)
UV transmittance ≥94.0 % (c=5, H2O, 400 nm) 1 cm cell
pH 11.1 to 12.0 (10 % in water)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR