Tisk…

Cyclophosphamide monohydrate 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-(Bis(2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide hydrate, Cyclophosphamide hydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-1,3,2-oxazaphosphorin-2-oxide

203960010 203960050 203960250
ACRO203960010EA 2470 CZK
ACRO203960010 ACRO203960050 ACRO203960250
Cyclophosphamide monohydrate 97%
Cyclophosphamide monohydrate

Appearance: White to off-white Crystalline powder or crystals

Vzorec: C₇H₁₇Cl₂N₂O₃P
Bod tání: 49…53 °C
Bod vzplanutí: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149395
CAS číslo: 6055-19-2
UN: 3464
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 53 °C
Water 5.5 to 7.5 % (K.F.)
HPLC ≥96.0 % (ELSD)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR