Tisk…

Amberlyst® A-21 ion-exchange resin

Dodavatel: Thermo Scientific

Synonyms: Amberlyst® A21

202170010 202172500
ACRO202170010EA 5960 CZK
ACRO202170010 ACRO202172500
Amberlyst® A-21 ion-exchange resin
Amberlyst® A-21

Appearance: Light brown =<1.80

Storage Temperature: Ambient
MDL Number: MFCD00145842
CAS číslo: 9049-93-8

Objednat

Výsledky specifikačního testu

Appearance Light brown Beads
Particle size ≤2.0 % (> 1.180 mm)
Harmonic mean size 0.490 to 0.690 mm
Uniformity coefficient ≤1.80
Moisture holding capacity 52.0 to 62.0 % (free base)
Total volume capacity ≥1.3 eq/l (free base)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR