Tisk…

Sulphamethazine 99%

Dodavatel: Thermo Scientific

Synonyms: 4,6-Dimethylsulphadiazine, Sulphadimidine, 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulphonamide

199300250 199301000
ACRO199300250EA 1340 CZK
ACRO199300250 ACRO199301000
Sulphamethazine 99%
Sulphamethazine

Appearance: White to light beige Crystalline powder

Vzorec: C₁₂H₁₄N₄O₂S
MW: 278,33 g/mol
Bod tání: 197…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00006066
CAS číslo: 57-68-1

Objednat

Výsledky specifikačního testu

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 200 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR