Polyphosphoric acid (≥84% P₂O₅), pure
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
196950010
196950025
196950010.EA
3230
CZK
196950010.
196950025.
Polyphosphoric acid (≥84% P₂O₅), pure
Polyphosphoric acid
Vzorec:
H(PO₃H)nOH Bod varu: 550 °C (760 mmHg) Hustota: 2.05 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00084480 CAS číslo: 8017-16-1 UN: 3264 ADR: 8 III |
Výsledky specifikačního testu
Appearance | Clear viscous liquid (May darken on storage) |
Color scale | ≤50 APHA |
Titration with NaOH | ≥82 % (as P2O5) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...