Rhodium 5% on activated carbon, dry, powder, unreduced
Dodavatel: Thermo Fisher Scientific
195440050
195440250
195440010
ACRO195440050EA
24310
CZK
ACRO195440050
ACRO195440250
ACRO195440010
Rhodium 5% on activated carbon, dry, powder, unreduced
Rhodium
Vzorec:
Rh MW: 102.9 Bod varu: 3400…3700 °C (1013 hPa) Bod tání: 1960 Hustota: 12.41 Storage Temperature: Ambient |
MDL Number:
MFCD00011201 CAS číslo: 7440-16-6 Merck Index: 12,08353 |
Výsledky specifikačního testu
Appearance | Grey-black to black Powder |
Assay | 4.5 to 5.5 % (Rh) |
Particle Size/Sieve analysis | <128 um: 99% |
Additional info |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...