Tisk…

Tetraethylammonium acetate tetrahydrate 97+%

Dodavatel: Thermo Scientific
Nebezpečí

194940250 194941000 194945000
ACRO194940250EA 1990 CZK
ACRO194940250 ACRO194941000 ACRO194945000
Tetraethylammonium acetate tetrahydrate 97+%
Tetraethylammonium acetate tetrahydrate

Appearance: Colorless to white Crystalline powder or crystals

Vzorec: C₁₀H₃₁NO₆
MW: 261,359 g/mol
Bod tání: 40…45 °C
Bod vzplanutí: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00149993
CAS číslo: 67533-12-4
UN: 1325
ADR: 4.1,II

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 46 °C
Titration with HClO4 97.0 to 103.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR