Tisk…
Poly(methyl methacrylate), M.W. ∼15000 (GPC)
Dodavatel: Thermo Fisher Scientific
Synonyms: Methacrylic Acid Methyl Ester Polymer, Methyl Methacrylate Polymer, PMMA
190690500 190691000
ACRO190690500EA 4890 CZK
ACRO190690500 ACRO190691000
Poly(methyl methacrylate), M.W. ∼15000 (GPC)
Poly(methyl methacrylate)
Appearance: White Beads
Vzorec: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
Bod vzplanutí: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00134349
CAS číslo: 9011-14-7
Objednat
Výsledky specifikačního testu
Appearance White Beads
Infrared spectrum Conforms
Loss on drying ≤3 %
Additional info bead size (typically): 10 to 100 micron