Tisk…

Phenylpyruvic acid sodium salt monohydrate 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-Oxo-3-phenylpropionic acid sodium salt monohydrate, Sodium phenylpyruvate monohydrate, Sodium 2-oxo-3-phenylpropanoate monohydrate

189730050 189730250
ACRO189730050EA 1750 CZK
ACRO189730050 ACRO189730250
Phenylpyruvic acid sodium salt monohydrate 98%
Phenylpyruvic acid sodium salt monohydrate

Appearance: White to light yellow Powder

Vzorec: C₉H₇NaO₃·H₂O
Bod tání: 260 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149109
CAS číslo: 122049-54-1

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water 7 to 12 % (K.F.)
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR