Tisk…

Sodium-1,2,4-triazolide 90%, Technical Grade

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1,2,4-Triazole, sodium derivative, 1,2,4-Triazole Sodium Salt, 1,2,4-Triazolylsodium, Sodium 1,2,4-triazol-1-ide, 1H-1,2,4-Triazole sodium salt

188990500 188992500 188990100
ACRO188990500EA 5960 CZK
ACRO188990500 ACRO188992500 ACRO188990100
Sodium-1,2,4-triazolide 90%, Technical Grade
Sodium-1,2,4-triazolide

CAS No.: 41253-21-8

Vzorec: C₂H₃N₃
Storage Temperature: Ambient
MDL Number: MFCD00044437
CAS číslo: 41253-21-8

Objednat

Výsledky specifikačního testu

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Identification (sodium): passes test
Titration with HClO4 ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR