Polyvinylacetate, M.W. ∼ 170,000
Dodavatel: Thermo Fisher Scientific
Synonyms:
PVA
183250010
183250250
183255000
ACRO183250010EA
9640
CZK
ACRO183250010
ACRO183250250
ACRO183255000
Polyvinylacetate, M.W. ∼ 170,000
Polyvinylacetate
Vzorec:
[CH₂CH(O₂CCH₃)]n Hustota: 1.18 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00084457 CAS číslo: 9003-20-7 |
Výsledky specifikačního testu
Appearance | Clear Beads |
Infrared spectrum | Conforms |
Loss on drying | ≤4 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...