Poly(caprolactone)diol, M.W. ∼ 2000
Dodavatel: Thermo Scientific
183212500
183215000
183210025
ACRO183212500EA
6140
CZK
ACRO183212500
ACRO183215000
ACRO183210025
Poly(caprolactone)diol, M.W. ∼ 2000
Poly(caprolactone)diol
CAS No.: 36890-68-3
Hustota:
1,07 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD23713083 CAS číslo: 36890-68-3 |
Výsledky specifikačního testu
Appearance | White Waxy solid |
Infrared spectrum | Conforms |
Water | ≤2 % (K.F.) |
Molecular number | 1500 to 2500 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...