Tisk…

Poly(N-vinylcarbazole), M.W. ∼90000

Dodavatel: Thermo Fisher Scientific

179030100 179030250
ACRO179030100EA 4560 CZK
ACRO179030100 ACRO179030250
Poly(N-vinylcarbazole), M.W. ∼90000
Poly(N-vinylcarbazole)

Appearance: White to cream Powder

Hustota: 1,2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00134336
CAS číslo: 25067-59-8

Objednat

Výsledky specifikačního testu

Appearance White to cream Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR