Tisk…

Polycarbonate resin, M.W. 45000, pellets

Dodavatel: Thermo Scientific

Synonyms: Poly(bisphenol-A-carbonate)

178310050 178312500 178315000
ACRO178310050EA 798 CZK
ACRO178310050 ACRO178312500 ACRO178315000
Polycarbonate resin, M.W. 45000, pellets
Polycarbonate resin

Appearance: Clear Pellets

MW: 45,000 g/mol
Hustota: 1,2 g/cm³
Bod vzplanutí: 449 °C
Storage Temperature: Ambient
MDL Number: MFCD00084476
CAS číslo: 24936-68-3

Objednat

Výsledky specifikačního testu

Appearance Clear Pellets
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR