Tisk…

N,N-Diethylaniline borane 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Borane - N,N-Diethylaniline Complex

176770250 176771000 176772500
ACRO176770250EA 2070 CZK
ACRO176770250 ACRO176771000 ACRO176772500
N,N-Diethylaniline borane 97%
N,N-Diethylaniline borane

Appearance: Clear colorless to amber Liquid

Vzorec: C₁₀H₁₈BN
MW: 163,07 g/mol
Bod tání: 30…27 °C
Hustota: 0,92 g/cm³
Bod vzplanutí: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00013187
CAS číslo: 13289-97-9
UN: 3148
ADR: 4.3,I

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5250 (20°C, 589 nm)
Titration ≥96.0 % (Active hydrogen)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR