Tisk…

Dichloromethylenedimethyliminium chloride 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

172210050 172210250 172211000
ACRO172210050EA 1340 CZK
ACRO172210050 ACRO172210250 ACRO172211000
Dichloromethylenedimethyliminium chloride 98%
Dichloromethylenedimethyliminium chloride

Appearance: White to light yellow Powder or crystals

Vzorec: C₃H₆Cl₃N
Bod tání: 181…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011870
CAS číslo: 33842-02-3
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
HPLC ≥97.5 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR