Tisk…

1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 99%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1-O-Acetyl-2,3,5-Tri-O-benzoyl-beta-D-Ribofuranose, (2S,3R,4R,5R)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate, β-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate, 2,3,5-Tri-o-Benzoyl-1-o-Acetyl-β-D-Ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate

170110100 170110250
ACRO170110100EA 4230 CZK
ACRO170110100 ACRO170110250
1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 99%
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose

CAS No.: 6974-32-9

Vzorec: C₂₈H₂₄O₉
Bod tání: 128…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005357
CAS číslo: 6974-32-9

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Heavy metals ≤20 ppm
HPLC ≥98.5 %
Specific optical rotation +22.0° to +26.0° (20°C, 589 nm) (c=1, pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR