Tisk…

O-Methyl-L-tyrosine 98%

Dodavatel: Thermo Scientific

Synonyms: (S)-2-Amino-3-(4-methoxyphenyl)propionic Acid, H-Tyr(Me)-OH, 4-Methoxy-L-phenylalanine

169750010 169752500
ACRO169750010EA 3080 CZK
ACRO169750010 ACRO169752500
O-Methyl-L-tyrosine 98%
O-Methyl-L-tyrosine

CAS No.: 6230-11-1

Vzorec: C₁₀H₁₃NO₃
MW: 195,22 g/mol
Bod tání: 259…261 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00002604
CAS číslo: 6230-11-1

Objednat

Výsledky specifikačního testu

Appearance White to light beige Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -7° to -9° (20°C, 589 nm) (c=2, 1 N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR