Tisk…

Quinidine, anhydrous 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (+)-Quinidine

163680250 163681000
ACRO163680250EA 6330 CZK
ACRO163680250 ACRO163681000
Quinidine, anhydrous 98%
Quinidine

CAS No.: 56-54-2

Vzorec: C20H24N2O2
MW: 324,42 g/mol
Bod tání: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00135581
CAS číslo: 56-54-2
UN: 1544
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 170 to 175 °C
Additional info Remainder dihydroquinidine
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR