Tisk…

Quinoline-N-oxide hydrate 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Quinoline 1-oxide hydrate

153430050 153430250
ACRO153430050EA 1430 CZK
ACRO153430050 ACRO153430250
Quinoline-N-oxide hydrate 98%
Quinoline-N-oxide hydrate

CAS No.: 1613-37-2

Vzorec: C₉H₇NO·xH₂O
Bod tání: 48…53 °C
Bod vzplanutí: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00149472
CAS číslo: 198878-42-1

Objednat

Výsledky specifikačního testu

Appearance Ochre to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 48 to 53 °C
Water ≤22.5 % (K.F.)
Titration with HClO4 ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR