Tisk…

(S)-2-Hydroxy-3-phenylpropanoic acid 98%

Dodavatel: Thermo Scientific

Synonyms: L-β-Phenyllactic acid, (S)-(-)-3-Phenyllactic Acid, L-3-Phenyllactic Acid, L(-)-3-Phenyllactic acid, (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, L-(-)-3-Phenyllactic acid

150920010 150920050 150920250
ACRO150920010EA 1020 CZK
ACRO150920010 ACRO150920050 ACRO150920250
(S)-2-Hydroxy-3-phenylpropanoic acid 98%
(S)-2-Hydroxy-3-phenylpropanoic acid

CAS No.: 20312-36-1

Vzorec: C₉H₁₀O₃
MW: 166,17 g/mol
Bod tání: 121…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004244
CAS číslo: 20312-36-1

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder or needles
Infrared spectrum Conforms
Melting point 121 to 125 °C
TLC ≥97.5 %
Specific optical rotation -17° to -20° (24 °C, 589 nm) (c=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR