L-serine methyl ester hydrochloride 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
H-Ser-OMe.HCl
132670050
132670250
132671000
ACRO132670050EA
1460
CZK
ACRO132670050
ACRO132670250
ACRO132671000
L-serine methyl ester hydrochloride 98%
L-serine methyl ester hydrochloride
CAS No.: 5680-80-8
Vzorec:
C₄H₉NO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00063680 CAS číslo: 5680-80-8 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Water | ≤1.0 % (K.F.) |
Sulfated ash | ≤0.1 % |
Appearance of solution | (5% in methanol) clear colorless to almost colorless |
Specific optical rotation | +3.5° to +5.5° (20°C, 589 nm) (c=2, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...