Oxytetracycline dihydrate 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
123841000
123840100
ACRO123841000EA
23090
CZK
ACRO123841000
ACRO123840100
Oxytetracycline dihydrate 95%
Oxytetracycline dihydrate
CAS No.: 6153-64-6
Vzorec:
C₂₂H₂₄N₂O₉·2H₂O Bod tání: 181…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151234 CAS číslo: 6153-64-6 |
Výsledky specifikačního testu
Appearance | Yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Water | 6 to 9 % (K.F.) |
HPLC | 94 to 102 % (on anhydrous substance) |
Specific optical rotation | on anhydrous substance |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...