4-Phenylpiperidine-4-carboxylic acid-4-toluenesulphonate 98%
Dodavatel: Thermo Scientific
|
Varování
|
122920250
122921000
ACRO122920250EA
1130
CZK
ACRO122920250
ACRO122921000
4-Phenylpiperidine-4-carboxylic acid-4-toluenesulphonate 98%
4-Phenylpiperidine-4-carboxylic acid-4-toluenesulphonate
Appearance: Off-white to light beige Crystalline powder
Vzorec:
C₁₂H₁₅NO₂·C₇H₈O₃S Bod tání: 225…229 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044709 CAS číslo: 83949-32-0 |
Výsledky specifikačního testu
Appearance | Off-white to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 225 to 229 °C |
Water | ≤1 % (K.F.) |
Solubility | (3 % in boiling water) Clear colorless to almost colorless |
Titration with NaOH | 97.5 to 102.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...