2',3'-O-Isopropylideneadenosine 98%
Dodavatel: Thermo Scientific
Synonyms:
2',3'-o-Isopropylideneadenosine
122720050
122720250
ACRO122720050EA
2110
CZK
ACRO122720050
ACRO122720250
2',3'-O-Isopropylideneadenosine 98%
2',3'-O-Isopropylideneadenosine
Appearance: White to off-white Crystalline powder
Vzorec:
C₁₃H₁₇N₅O₄ MW: 307,31 g/mol Bod tání: 221…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005756 CAS číslo: 362-75-4 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
UV spectrophotometry | molar absorptivity 15400 +/-4%, on dry substance |
Ratio of Absorbances | ≤0.01 (A290/A260) |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...