Tisk…

Isochroman 99%

Dodavatel: Thermo Fisher Scientific

122540250 122540050
ACRO122540250EA 2340 CZK
ACRO122540250 ACRO122540050
Isochroman 99%
Isochroman

Appearance: Clear colorless to yellow Liquid

Vzorec: C₉H₁₀O
Bod varu: 75 °C (4mbar)
Hustota: 1,06 g/cm³
Bod vzplanutí: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006913
CAS číslo: 493-05-0

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5460 (20°C, 589 nm)
GC ≥98.5 %
Titration with NaOH ≥98.5 % (On anhydrous basis)
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR