D-(+)-Glucosamine hydrochloride 98+%
Dodavatel: Thermo Scientific
Synonyms:
2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
119900100
119901000
119905000
119900025
ACRO119900100EA
1130
CZK
ACRO119900100
ACRO119901000
ACRO119905000
ACRO119900025
D-(+)-Glucosamine hydrochloride 98+%
D-(+)-Glucosamine hydrochloride
Appearance: White Crystalline powder
Vzorec:
C₆H₁₃NO₅ MW: 215,63 g/mol Bod tání: 190…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135831 CAS číslo: 66-84-2 |
Výsledky specifikačního testu
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1.0 % (105°C, 2 hrs) |
Sulfated ash | ≤0.1 % |
Specific optical rotation | +70.0° to +73.0° (20°C, 589 nm) (c=2.5, H2O) (after 3 h) |
Titration with HClO4 | 98.0 to 102.0 % (On dry substance) |
Arsenic (As) | ≤3 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...